EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
A convenient one-step synthesis of L-aminotryptophans and improved synthesis of 5-fluorotryptophan.
PMID 18667314
Abstract
A one-pot biotransformation for the generation of a series of L-aminotryptophans using a readily prepared protein extract containing tryptophan synthase is reported. The extract exhibits remarkable stability upon freeze-drying, and may be stored and used for long periods after its preparation without significant loss of activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0896
5-Fluoro-DL-tryptophan, powder
C11H11FN2O2
47568
5-Fluoro-L-tryptophan, ≥98.0% (HPLC)
C11H11FN2O2