EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Ketoisophorone transformation by Marchantia polymorpha and Nicotiana tabacum cultured cells.
PMID 18669027
Abstract
Stereospecific olefin (C=C) and carbonyl (C=O) reduction of the readily available prochiral compound ketoisophorone (2,2,6-trimethyl-2-cyclohexene-1,4-dione) (1) by Marchantia polymorpha and Nicotiana tabacum cell suspension cultures produce the chiral products (6R)-levodione (2), (4R,5S)-4-hydroxy-3,3,5-trimethylcyclohexanone (3), and (4R,6R)-actinol (4) as well as the minor components (4R)-hydroxyisophorone (5) and (4S)-phorenol (6).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329517
2,6,6-Trimethyl-2-cyclohexene-1,4-dione, 98%
C9H12O2
W342106
4-Oxoisophorone, ≥98%, FG
C9H12O2