EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Functionalizing glycine derivatives by direct C-C bond formation.
PMID 18671311
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A16300
N-Acetylglycine, ReagentPlus®, 99%
C4H7NO3