EMAIL THIS PAGE TO A FRIEND
The American journal of medicine
Hypercholesterolemia: new findings and clinical applications. Introduction.
PMID 1867229
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5200
Triparanol, ≥97% (HPLC), powder
C27H32NO2Cl