EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Transformation of N-phenylpiperazine by mixed cultures from a municipal wastewater treatment plant.
PMID 18676696
Abstract
Samples from a wastewater treatment plant were used as inocula for mixed cultures dosed with N-phenylpiperazine (NPP), a model compound containing the piperazine ring found in many fluoroquinolones. Chemical analyses showed that NPP (50 mg liter(-1)) disappeared in 12 days, with the appearance of a transient metabolite and two nitrosated compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P30004
1-Phenylpiperazine, 99%
C10H14N2