EMAIL THIS PAGE TO A FRIEND
Beijing da xue xue bao. Yi xue ban = Journal of Peking University. Health sciences
[Synthesis of diethyl N-[p-(methylamino)benzoyl]-L-glutamate].
PMID 18677396
Abstract
To find a best synthesis method of diethyl N-[p-(methylamino)benzoyl]-L-glutamate(1). Using diethyl N-(p- aminobenzoyl)-L-glutamate (2) as a starting material via a one-pot process of benzylation and methylation, the diethyl N-[p-(methylamino)benzoyl ]-L-glutamate (3) is prepared in high yield. Then compound 1 was conveniently obtained from debenzylation of compound 3 by Pd/C. The target compound was obtained through 2-step reaction in 88% overall yield in only 6 h, and its structure was identified by 1HNMR and MS. It is a convenient and efficient method of preparation of diethyl N-[p-(methylamino)benzoyl]-L-glutamate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
309346
L-Glutamic acid diethyl ester hydrochloride, 97%
C9H17NO4 · HCl