EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Directed assembly of sub-nanometer thin organic materials with programmed-size nanopores.
PMID 18677728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2354
α-D(+)-Glucose pentaacetate, 99%
C16H22O11
285943
β-D-Glucose pentaacetate, 98%
C16H22O11