EMAIL THIS PAGE TO A FRIEND
International journal of environmental research and public health
Juvenile Hormone Analogues, methoprene and fenoxycarb dose-dependently enhance certain enzyme activities in the silkworm Bombyx mori (L).
PMID 18678927
Abstract
Use of Juvenile Hormone Analogues (JHA) in sericulture practices has been shown to boost good cocoon yield; their effect has been determined to be dose-dependent. We studied the impact of low doses of JHA compounds such as methoprene and fenoxycarb on selected key enzymatic activities of the silkworm Bombyx mori. Methoprene and fenoxycarb at doses of 1.0 microg and 3.0 fg/larvae/48 hours showed enhancement of the 5th instar B. mori larval muscle and silkgland protease, aspartate aminotransaminase (AAT) and alanine aminotransaminase (ALAT), adenosine triphosphate synthase (ATPase) and cytochrome-c-oxidase (CCO) activity levels, indicating an upsurge in the overall oxidative metabolism of the B.mori larval tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34343
Fenoxycarb, PESTANAL®, analytical standard
C17H19NO4