EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Selective structure-based virtual screening for full and partial agonists of the beta2 adrenergic receptor.
PMID 18680279
Abstract
The recently solved high-resolution X-ray structure of the beta2 adrenergic receptor has been challenged for its ability to discriminate inverse agonists/antagonists from partial/full agonists. Whereas the X-ray structure of the ground state receptor was unsuitable to distinguish true ligands with different functional effects, modifying this structure to reflect early conformational events in receptor activation led to a receptor model able to selectively retrieve full and partial agonists by structure-based virtual screening. The use of a topological scoring function based on molecular interaction fingerprints was shown to be mandatory to properly rank docking poses and achieve acceptable enrichments for partial and full agonists only.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53787
Carazolol, analytical standard
C18H22N2O2