EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Versatile three-component procedure for combinatorial synthesis of 2-aminospiro[(3'H)-indol-3',4-(4H)-pyrans].
PMID 18680348
Abstract
A convenient method of synthesis of substituted and annulated 2-amino-spiro[(3' H)-indol-3',4-(4 H)-pyrans] at mild conditions and in good yields is developed. Three component reaction of wide variety of substituted isatins, cyanoacetic acid derivatives, and carbonyl compounds or phenols gives the target compounds. Forty new spiropyrans were obtained, and their structures were proved by elemental analysis and 1H NMR and IR spectral data. It is shown that the use of a not very large set of starting compounds can lead to the synthesis of a thousand-member 2-amino-spiro[(3' H)-indol-3',4-(4 H)-pyran] library.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C88505
Cyanoacetic acid, 99%
C3H3NO2