EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Microelectrode arrays and ceric ammonium nitrate: a simple strategy for developing new site-selective synthetic methods.
PMID 18680369
Abstract
Conditions for a site-selective ceric ammonium nitrate oxidation have been developed. The reactions proceed nicely on both 1K- and 12K-microelectrode arrays. The procedure for developing the reactions was very simple and demonstrated that the same reagents used for a solution-phase reaction can be used for a related site-selective reaction on a microelectrode array.