EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Human adenosine deaminase as an allosteric modulator of human A(1) adenosine receptor: abolishment of negative cooperativity for [H](R)-pia binding to the caudate nucleus.
PMID 18680557
Abstract
It has been shown that adenosine deaminase (ADA; EC 3.5.4.4) behaves as an ecto-enzyme anchored to membrane proteins, among them A(1) adenosine receptors (A(1)Rs). Bovine ADA interacts with A(1)Rs from many species and regulates agonists binding to receptors in an activity-independent form. However, it was not known whether human ADA exerted any effect on the agonist binding to human A(1)Rs, because of both technical difficulties in obtaining pure human ADA and tissues containing human A(1)Rs. In this study, human ADA was purified to homogeneity. Taking in consideration that A(1)Rs form homodimers and taking advantage of a new procedure to fit binding data to receptors dimers, which allows to calculate ligand dissociation constants and the degree of cooperativity between the two subunits in the dimer, here it is demonstrated that human ADA markedly enhances the agonist and antagonist affinity and abolishes the negative cooperativity on agonist binding to human striatal A(1)Rs. ADA also increases the ability of the agonist to decrease the forskolin-induced cAMP levels. The results show that human ADA, apart from reducing the adenosine concentration and thus preventing A(1)R desensitization, binds to A(1)R behaving as an allosteric effector that markedly enhances agonist affinity and increases receptor functionality. The physiological role of the interaction is to make receptors more sensitive to adenosine. This powerful regulation has important implications for the physiology and pharmacology of neuronal A(1)Rs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4532
(−)-N6-(2-Phenylisopropyl)adenosine, solid
C19H23N5O4