EMAIL THIS PAGE TO A FRIEND
Blood
Inhibition of human platelet reactivity by endothelium-derived relaxing factor from human umbilical vein endothelial cells in suspension: blockade of aggregation and secretion by an aspirin-insensitive mechanism.
PMID 1868238
Abstract
To determine a role for endothelium-derived relaxing factor/nitric oxide (EDRF/NO) in regulation of human platelet reactivity by human endothelial cells (EC), we studied combined suspensions of human umbilical vein endothelial cells (HU-VEC, passage 2 through 3) and washed human platelets. Confluent HUVEC monolayers were treated with aspirin (1 mmol/L) to prevent prostacyclin (PGI2) formation, washed, and harvested. Aspirin-treated platelets alone (58 x 10(6)) were fully aggregated by thrombin at 0.05 U/mL or more. In the presence of 10(6) HUVEC, however, platelet serotonin release and aggregation in response to thrombin at doses as high as 0.5 U/mL were blocked. We demonstrated for the first time that inhibition of aggregation and serotonin release, due to EDRF/NO, occurred in parallel. HUVEC-dependent inhibition of platelet responsiveness was enhanced by superoxide dismutase (SOD) and reversed by hemoglobin. The inhibitory effect was also reversed by preincubation of HUVEC with NG-monomethyl-L-arginine (NMA) or NG-nitro-L-arginine (NNA) through competitive blockade of arginine metabolism. Pretreatment of platelets with methylene blue indicated that EC-dependent inhibition of platelet reactivity occurred through activation of platelet soluble guanylate cyclase. When platelets and HUVEC were separated by a permeable membrane and both cells were stimulated by thrombin, platelets remained unresponsive. This indicated that inhibition was induced by a fluid-phase mediator, independent of direct cell-cell contact. These data demonstrate that EDRF/NO formation from L-arginine by human EC plays an important role as an aspirin-insensitive fluid-phase inhibitor of human platelet reactivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7379 Hemoglobin human, lyophilized powder