EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
FOXA transcription factors determine the amplitude of glucocorticoid induction of tyrosine aminotransferase in mice.
PMID 18683506
Abstract
o-Aminoazotoluene was more potent than 3'-methyl-4-dimethylaminoazobenzene in modulating glucocorticoid induction of tyrosine aminotransferase and DNA-binding activity of FOXA (HNF3) in 12-day-old ICR mice. In adult animals, induction of tyrosine aminotransferase and FOXA activity were modulated by o-aminoazotoluene, while 3'-methyl-4-dimethylaminoazobenzene was ineffective. Our results suggest that FOXA proteins determine glucocorticoid induction of tyrosine aminotransferase in mice (similarly to rats).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
121568
Fast Garnet GBC base, 97%
C14H15N3
31629
o-Aminoazotoluene, analytical standard
C14H15N3