EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Stereoselective C9 carbon-carbon couplings of quinine: synthesis and conformational analysis of new C2-symmetric dimers.
PMID 18687001
Abstract
An unexpected stereoselective direct dimerization occurred when 9-quinine halide was treated with butyllithium. The reaction of either (9S)- or (9R)-chloroquinine gave the same C2-symmetric dimer with 9R configuration (X-ray structure). A tentative mechanism involving radical recombination is discussed. This highly congested dimer forms two atropisomers, and their reversible interconversion was studied by NMR. Another C2-symmetric (9S)-quinine dimer connected solely by carbon-carbon bonds was obtained by the stereoselective coupling of bis(arylbromomagnesium) derivative with (9S)-chloroquinine.