EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Fluorination reactions in microreactors.
PMID 18688335
Abstract
The DAST-mediated conversion of a range of alcohols to the corresponding fluorides in a microstructured device is described. This safe, practical fluorination method will facilitate reactions currently challenging on large scale.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235253
(Diethylamino)sulfur trifluoride, 95%
C4H10F3NS