EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
In vitro and in vivo evaluations of oxacillin efficiency against mecA-positive oxacillin-susceptible Staphylococcus aureus.
PMID 18694946
Abstract
Community-type Staphylococcus aureus strains that are positive for mecA and PBP2a but appear phenotypically susceptible to oxacillin are increasingly reported worldwide. Four S. aureus clinical isolates carrying the mecA gene with oxacillin MICs of <2 microg/ml were tested for oxacillin efficiency by population analyses and experimental thigh infections. These isolates harbored staphylococcal cassette chromosome mec type IV and belonged to two genotypes. Two of the four isolates were found by population analysis to be truly oxacillin susceptible. All four isolates exhibited significant reductions in the numbers of colonies grown after dicloxacillin treatment of experimental thigh infections, as also did a mecA-negative S. aureus control strain. These observations indicate that some of the phenotypically oxacillin susceptible mecA-positive Staphylococcus aureus isolates may be at least partially responsive to oxacillin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9016
Dicloxacillin sodium salt monohydrate
C19H16Cl2N3NaO5S·H2O