EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
Characterization of beta-D-xyloside-induced glycosaminoglycans and oligosaccharides in cultured human skin fibroblasts.
PMID 1869505
Abstract
Human skin fibroblasts were incubated in the presence of a fluorogenic xyloside, 4-methylumbelliferyl beta-D-xyloside. Three fluorogenic components were isolated and purified from the culture medium by gel permeation high-performance liquid chromatography. Their structures were then characterized by enzymatic digestion, fast-atom-bombardment mass spectrometry, gas-liquid chromatography, and electrophoresis on cellulose acetate membrane. The results showed that one of the components was a mixture of dermatan sulfate (70%) and chondroitin sulfate (30%), bearing the 4-methylumbelliferone at the reducing termini, and having an average molecular weight of 9,200. The others had the structures galactosyl-galactosyl-xylosyl-4-methylumbelliferone and galactosyl-xylosyl-4-methylumbelliferone, respectively, representing the linkage region between the glycosaminoglycan chains and core protein, except that 4-methylumbelliferone replaced the amino acid. Moreover, it was demonstrated that these oligosaccharides were intermediates of glycosaminoglycan synthesis, not depolymerized products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7008
4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate
C15H16O7