EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[New promising antioxidants based on 2,6-dimethylphenol].
PMID 18695730
Abstract
Three new sulfur-containing derivatives of 2,6-dimethylphenol were synthesized. Their antioxidative activity, mutagenicity, and genotoxicity were examined by bacterial tests and by calculating the dominant lethal mutations in murine embryonic cells. It was shown that all the compounds synthesized have a marked antioxidative effect and no genotoxic or mutagenic properties. One of the antioxidants, 4-(3-dodecylthiopropyl)-2,6-dimethylphenol, increases the survival of cells of both the wild-type Escherichia coli strain and bacterial strains defective in the genes of repair enzymes and has a more distinct antioxidative effect than the classic antioxidants alpha-tocopherol and trolox, increasing the survival of cells devoid of repair enzymes.