EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
Induction of apoptosis by the inhibitors of poly(ADP-ribose)polymerase in HeLa cells.
PMID 18695944
Abstract
To investigate the role of poly(ADP-ribose)polymerase (PARP) in the physiological condition of cell growth, we studied the ability of PARP inhibitors to induce apoptosis. Benzamide (BA) and 4-amino-1,8-naphthalimide (NAP), two well-known inhibitors of PARP, treatment increased nuclear fragmentation and caspase-3 activity in HeLa (Human cervical cancer cell line) cells. The increase of cellular NAD(+) level was observed in HeLa cells treated with BA in comparison with untreated control cells. For unrevealing the specific PARP family member responsible for such induction of apoptosis we knocked down and over-expressed PARP-1 gene in HeLa cells. PARP-1 knock down cells were sensitive to BA induced nuclear fragmentation and caspase-3 activation while exogenous expression of PARP-1 rendered cells resistant to BA induced apoptosis. This result indicated that inhibition of PARP-1 resulted in induction of apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0966
4-Amino-1,8-naphthalimide
C12H8N2O2