EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
De novo synthesis of (+)-isofregenedol.
PMID 18698826
Abstract
An efficient enantioselective synthesis of (+)-isofregenedol was achieved in 13 steps from commercially available cyclohexene oxide without the use of protecting groups. The tetrahydronaphthalenic core of isofregenedol was obtained via a gold(I)-catalyzed benzannulation recently developed in our laboratory.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102504
Cyclohexene oxide, 98%
C6H10O