EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
let-7 regulates Dicer expression and constitutes a negative feedback loop.
PMID 18700235
Abstract
microRNAs (miRNA) are small, endogenously expressed non-coding RNAs that are sequentially processed by Drosha and Dicer from primary transcripts, by negatively regulating the expression of protein-coding genes through either translational repression or RNA degradation. Their expression patterns are developmentally regulated and/or tissue specific, while altered expressions of certain miRNAs are frequently observed in human cancers, though the underlying regulatory mechanism is largely unknown. Herein, we show that Dicer expression was inversely correlated with expression levels of mature let-7 in a panel of human cancer cell lines, showing association with cell growth and cell cycle phases. Overexpression of let-7 significantly reduced the expression of Dicer at both the protein and messenger RNA levels, whereas antisense-mediated reduction of let-7 expression conversely increased Dicer at both levels. A luciferase assay using a reporter carrying a putative target site in the 3' untranslated region of Dicer revealed that let-7 directly affects Dicer expression. Downregulation of Dicer resulted in a reduced expression of mature let-7. Furthermore, overexpression of let-7 decreased the levels of expression of other mature miRNAs, while knockdown of let-7 increased those levels. Taken together, these findings strongly suggest the possible existence of a novel regulatory loop, in which let-7 may play a role as a key miRNA for implementing the tightly regulated, equilibrated state of Dicer and various miRNAs.