EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and bio-evaluation of a new fatty acid derivative for myocardial imaging.
PMID 18701306
Abstract
Development of a (99m)Tc-fatty acid analogue is of interest, as (99m)Tc is logistically advantageous over the cyclotron-produced (11)C and (123)I. Synthesis of a 16 carbon fatty acid derivative and its radiolabeling with the novel [(99m)TcN(PNP)](2+) core is described here. Hexadecanedioic acid was conjugated to cysteine in an overall yield of 55%. This ligand could be labeled with (99m)Tc via the [(99m)TcN(PNP)](2+) core, in 80% yield, as a mixture of two isomers (syn and anti). The major isomer isolated by HPLC was used for bioevaluation studies in swiss mice and compared with radioiodinated iodophenyl pentadecanoic acid (IPPA), an established agent for myocardial metabolic imaging. (99m)Tc-labeled complex cleared faster from the non-target organs, namely, liver, lungs, and blood compared to that of [(125)I]-IPPA. However, the complex exhibited lower uptake and faster washout from the myocardium as compared to [(125)I]-IPPA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
177504
Hexadecanedioic acid, 96%
C16H30O4