EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of N-allylideneamines and their use for the double nucleophilic addition of ketene silyl (thio)acetals and trimethylsilyl cyanide.
PMID 18702497
Abstract
N-Allylideneamines 1a, b were prepared from acrolein and diphenylethyl or trityl amine in the presence of Ti(OEt)4. Double nucleophilic addition of various ketene silyl (thio)acetals and trimethylsilyl cyanide to these imines proceeded efficiently to give, after workup with TFA, homoglutamic acid derivatives 3 and valerolactam 5.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73328
Trimethylsilyl cyanide, technical, ≥95% (GC)
C4H9NSi
212849
Trimethylsilyl cyanide, 98%
C4H9NSi