EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Chemistry. Halogen versus hydrogen.
PMID 18703728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852473
5-Bromouracil, 98%
C4H3BrN2O2