EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Separation and characterization of oxaliplatin dinucleotides from DNA using HPLC-ESI ion trap mass spectrometry.
PMID 18709362
Abstract
Oxaliplatin is a third-generation platinum complex, and has a broad spectrum of antitumor activity. Such platinum complexes with the DACH carrier ligand have recently received increasing attention since they show efficacy against cisplatin-resistant cell lines. As the foremost indication of antitumor activity of platinum drugs is the formation of adducts with genomic DNA, calf thymus DNA-oxaliplatin adducts were the major target in this study. Calf thymus DNA was incubated with oxaliplatin, resulting in the formation of a large number of platinum-DNA adducts. Treated DNA was digested into the dinucleotides with a combination of enzymes, namely, benzonase, alkaline phosphatase, and nuclease S1. Using a high-performance liquid chromatography, we carried out the separation of individual platinum-DNA adducts which were concurrently identified using electrospray ionization ion trap mass spectrometry (MS). Both 1,2-intrastrand and 1,2-interstrand cross-linked adducts were found; however, those of the intrastrand nature have a considerably higher abundance than those of the interstrand cross-links. Among them, d(GpG)-oxaliplatin was the most abundant bifuctional adduct. To a lesser extent, a few monofunctional adducts were detected as well. MS(n) experiments served to ascertain the detailed structures of oxaliplatin adducts of dinucleoside monophosphates and of dinucleotides.