EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
N2-functionalized blue luminescent guanosines by 2,2'-dipyridylamino and 2-(2'-pyridyl)benzimidazolyl chelate groups and their interactions with Zn(II) ions.
PMID 18710219
Abstract
The syntheses of new blue luminescent N(2)-modified guanosine derivatives with chromophores p-4,4'-biphenyl-NPh2 (1a), p-4,4'-biphenyl-N(2-py)2 (1b), and p-4,4'-biphenyl-2-(2'-pyridyl)benzimidazolyl (1c), respectively, have been achieved. These new N(2)-guanosines are moderate blue emitters with lambda(max) = 395 nm (1a), 370 nm (1b), and 403 nm (1c) and Phi = 0.13, 0.07, and 0.10 in tetrahydrofuran, respectively. Spectroscopic studies and density-functional theory calculations established that the guanine moiety and the new chromophore in all three molecules are involved in the luminescent process. We have also established that guanosines 1a-1c can interact with metal ions such as Zn(II). The interactions of Zn(II) ions with the three guanosines were examined via absorption, fluorescence, circular dichroism (CD), and NMR spectroscopic analyses. We have found that these guanosines display a distinct fluorescent response toward Zn(II) ions which can be attributed to the presence of the chelate chromophore N(2-py)2 in 1b and 2-py-benzimidazolyl in 1c. For 1a and 1b, the addition of Zn(II) ions causes straight fluorescent quenching while for 1c the addition of Zn(II) ions causes quenching initially, which is followed by a distinct spectral red shift and the intensity enhancement of the new emission peak. NMR and CD studies demonstrated that the Zn(II) ions bind preferentially to the guanine moiety in 1a and 1b but to the 2-(2'-py)benzimidazolyl chelate site in 1c. Moreover, the anion-dependent CD response of 1a-1c toward Zn(II) salts points to the possible involvement of intramolecular hydrogen bonding between the acetate bound to the Zn(II) ion and the hydroxyl groups of the guanosine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D216402
2,2′-Dipyridylamine, 98%
C10H9N3