EMAIL THIS PAGE TO A FRIEND
Archives of dermatology
New contrast stain for the rapid diagnosis of pityriasis versicolor.
PMID 18711086
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8679
Chicago Sky Blue 6B, powder
C34H24N6Na4O16S4