EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Novel enamine derivatives of 5,6-dihydro-2'-deoxyuridine formed in reductive amination of 5-formyl-2'-deoxyuridine.
PMID 18711667
Abstract
Reductive amination of 5-formyl-3',5'-di-O-acetyl-2'-deoxyuridine with primary amines and sodium triacetoxyborohydride (NaBH(OAc)(3)) afforded novel enamine derivatives of 5,6-dihydro-2'-deoxyuridine as a result of unexpected 1,4-conjugate reduction of intermediate Schiff bases in addition to the secondary amine derivatives of 2'-deoxyuridine, typical 1,2-reduction products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471305
Butylamine, 99.5%
C4H11N
W313009
Butylamine, ≥99%
C4H11N