EMAIL THIS PAGE TO A FRIEND
Biomedica : revista del Instituto Nacional de Salud
[Evaluation of the triflumuron and the mixture of Bacillus thuringiensis plus Bacillus sphaericus for control of the immature stages of Aedes aegypti and Culex quinquefasciatus (Diptera: Culicidae) in catch basins].
PMID 18719724
Abstract
In Cali, Colombia, catch basins (streetside storm drains) are one of the main larval habitats of Aedes aegypti and Culex quinquefasciatus. Since 1999, these mosquitoes have been controlled by the Secretaría de Salud Municipal (Secretary of Municipal Public Health) using the larvicide triflumuron. Because of high densities of these mosquitoes that remain in the city, treatment failure was suspected -possibly insecticide resistance of the target species. The efficacy of triflumuron and VectoMax (biorational mixture of Bacillus thuringiensis var. israelensis plus Bacillus sphaericus) were evaluated in the control of A. aegypti and C. quinquefasciatus in catch basins. The residual effect of a single application of the biorational formulation was determined in catch basins during periods of high and low rainfall. The efficacy of the products was measured in 60 catch basins located in a residential neighborhood of Cali for a period of 90 days. The mean number of immature instars (A. aegypti and C. quinquefasciatus larvae and pupae of both species) was determined biweekly from 40 catch basins with insecticide intervention (20 treated with triflumuron, 20 with VectoMax) and 20 untreated (control group). The residual effect of the biorational larvicide was evaluated biweekly in 10 catch basins during each of the 2 climatic periods. Results. The catch basins treated with VectoMax presented a significantly lower mean number of immature instars of both species compared with the control ( p<0.01). In contrast, the triflumuron treatment significantly reduced only immature instars of A. aegypti compared with the control ( p<0.001). The residual effect of VectoMax was higher during low rainfall compared to the control ( p<0.001). Conclusion. The biorational formulation was the more effective treatment for the control of both species during the period of evaluation (15 days).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35029
Triflumuron, PESTANAL®, analytical standard
C15H10ClF3N2O3