EMAIL THIS PAGE TO A FRIEND
Cell Motility and the Cytoskeleton
Arp2 depletion inhibits sheet-like protrusions but not linear protrusions of fibroblasts and lymphocytes.
PMID 18720401
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52619
Hexamethyldisilazane, for GC derivatization, ≥99.0% (GC)
C6H19NSi2