EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Toward a full characterization of native starch: separation and detection by size-exclusion chromatography.
PMID 18722623
Abstract
The structure of starch molecules is relevant to nutrition and industrial applications. Size-exclusion chromatography (SEC, also known as GPC) of native starch generally suffers non-satisfactory repeatability and reproducibility of the dissolution and separation. This work combines two polar organic solvents: dimethylsulfoxide for complete dissolution and dimethylacetamide to limit shear degradation. The separation is as repeatable as that of polystyrene standards performing dissolution and separation at 80 degrees C. Successful covalent-labeling on the glucose unit is claimed to be published here for the first time in non-degradative conditions and allows the use of UV detector with significantly higher sensitivity than with a refractometer.