EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery and optimization of (R)-prolinol-derived agonists of the Growth Hormone Secretagogue receptor (GHSR).
PMID 18722770
Abstract
The discovery and optimization of a novel series of prolinol-derived GHSR agonists is described. This series emerged from a 11,520-member solid-phase library targeting the GPCR protein superfamily, and the rapid optimization of low micromolar hits into single-digit nanomolar leads can be attributed to the solid-phase synthesis of matrix libraries, which revealed multiple non-additive structure-activity relationships. In addition, the separation of potent diastereomers highlighted the influence of the alpha-methyl stereochemistry of the phenoxyacetamide sidechain on GHSR activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
186511
(S)-(+)-2-Pyrrolidinemethanol, 97%
C5H11NO