EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Chloroalkylsulfonate ionic liquids by ring opening of sultones with organic chloride salts.
PMID 18726017
Abstract
An efficient route to prepare ionic liquids with chloroalkylsulfonate anions is presented; the synthesis proceeds in a one-step ring-opening reaction of sultones with an organic chloride salt and provides a very attractive access to new anion functionalised ionic liquids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
316539
1,8-Naphthosultone, 98%
C10H6O3S