EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents of the pericarp of Trichosanthes rosthornii Harms].
PMID 1872972
Abstract
Nine compounds were isolated from the pericarp of T. rosthornii and identified on the basis of spectral data to be palmitic acid, lignoceric acid, cerotic acid, montanic acid, melissic acid, L-(-)-alpha-monopalmitin, delta 7-stigmastenol, delta 7-stigmastenone-3 and delta 7-stigmatenol-3-beta-D-glucopyranoside. All but the last one are obtained from this species for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1640
DL-α-Palmitin, ≥99%
C19H38O4