EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents of Osbeckia chinensis L].
PMID 1872973
Abstract
Five constituents, 2-furoic acid, succinic acid, ursolic acid, quercetin and daucosterol were isolated and identified from Osbeckia chinensis. All of them are reported to have been found from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F20505
2-Furoic acid, 98%
C5H4O3