EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Mercury vapor release from broken compact fluorescent lamps and in situ capture by new nanomaterial sorbents.
PMID 18754507
Abstract
The projected increase in the use of compact fluorescent lamps (CFLs) motivates the development of methods to manage consumer exposure to mercury and its environmental release at the end of lamp life. This work characterizes the time-resolved release of mercury vapor from broken CFLs and from underlying substrates after removal of glass fragments to simulate cleanup. In new lamps, mercury vapor is released gradually in amounts that reach 1.3 mg or 30% of the total lamp inventory after four days. Similar time profiles but smaller amounts are released from spent lamps or from underlying substrates. Nanoscale formulations of S, Se, Cu, Ni, Zn, Ag, and WS2 are evaluated for capture of Hg vapor under these conditions and compared to conventional microscale formulations. Adsorption capacities range over 7 orders of magnitude, from 0.005 (Zn micropowder) to 188 000 microg/g (unstabilized nano-Se), depending on sorbent chemistry and particle size. Nanosynthesis offers clear advantages for most sorbent chemistries. Unstabilized nano-selenium in two forms (dry powder and impregnated cloth) was successfully used in a proof-of-principle test for the in situ, real-time suppression of Hg vapor escape following CFL fracture.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A7511
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
A4161
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
A9430
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A2934
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
B6917
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
A3156
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
A9543
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A9056
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
A9085
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
A7517
Bovine Serum Albumin, For use as a marker in SDS-PAGE
A8654
Bovine Serum Albumin, dimer mol wt ~132 kDa
A3733
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
A3983
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
A3608
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
A3858
Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis)
A4737
Bovine Serum Albumin, lyophilized powder, protease, essentially free, ≥98% (agarose gel electrophoresis)
A4612
Bovine Serum Albumin, lyophilized powder, Essentially fatty acid free