EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Angiogenesis and growth factor modulation induced by alternagin C, a snake venom disintegrin-like, cysteine-rich protein on a rat skin wound model.
PMID 18755139
Abstract
In this work, we show that alternagin-C (ALT-C) and ALT-C PEP, a peptide derived from its sequence, were able to induce angiogenesis in wounded rat skin. A spherical cutaneous excision was made in the back of each animal and treated with three different concentrations of ALT-C or ALT-C PEP. After that, the skin was removed and analyzed to verify the presence of new vessels and the expression of growth factors. ALT-C and ALT-C PEP induced the formation of new vessels and modulated the expression of growth factors, mainly VEGF and FGF1. The expression of VEGF increased and it could be detected up to 7 days after injury. FGF1 also significantly increased, but at a lesser extent than VEGF. In conclusion, the present study shows for the first time the stimulation of angiogenesis in an injured tissue by a disintegrin-like protein and that ALT-C may exert this effect by modulating the expression of growth factors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5765
Prostaglandin F, synthetic
C20H36O5