EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Primary sensitization to toluene-2,5-diamine giving rise to early positive patch reaction to p-phenylenediamine and late to toluene-2,5-diamine.
PMID 18759912
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
154334
2,5-Diaminotoluene sulfate, 97%
C7H10N2 · H2SO4