EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Sedative activity of hexane extract of Keampferia galanga L. and its active compounds.
PMID 18761077
Abstract
It is well known that fragrance impacts behaviors and autonomic functions, and is increasingly used as relaxant, carminative, as well as sedative in aromatherapy. Kaempferia galanga L. is one of the popular traditional aromatic medicinal plants used in tropics and subtropics of Asia including China, Japan and Indochina. The aim of present study was to investigate sedative effects of hexane extract of Kaempferia galanga L.and 2 active aromatic compounds (compound 1: ethyl trans-p-methoxycinnamate and compound 2: ethyl cinnamate ) included in the extyract by means of inhalation in mice. Sedative activity was assessed by inhalation of the hexane extract and two major aromatic compounds. Inhalation of hexane extract at the doses of 1.5 and 10mg showed significant reduction of locomotor activity, indicating considerable sedative and relaxant effects. Compound 1 and 2 were proved to possess sedative effects at 0.0014mg and 0.0012mg respectively. These results suggest the promising application of Kaempferia galanga L. and its constituents in aromatherapy.
Related Materials