EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Palladium-catalyzed method for the synthesis of carbazoles via tandem C-H functionalization and C-N bond formation.
PMID 18761437
Abstract
The development of a new method for the assembly of unsymmetrical carbazoles is reported. The strategy involves the selective intramolecular functionalization of an arene C-H bond and the formation of a new arene C-N bond. The substitution pattern of the carbazole product can be controlled by the design of the biaryl amide substrate, and the method is compatible with a variety of functional groups. The utility of the new protocol was demonstrated by the concise synthesis of three natural products from commercially available materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
540633
2′-Bromoacetanilide, 96%
C8H8BrNO