EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Identification of 14-3-3zeta by chemical affinity with salicylanilide inhibitors of interleukin-12p40 production.
PMID 18763820
Abstract
Salicylanilides were found as selective inhibitors of interleukin-12p40 production in stimulated dendritic cells. The conversion of one of these bioactive salicylanilides into a comparably bioactive, chemically labeled derivative was achieved using a facile and systematic functional group derivatization strategy. This resulted in a tool reagent that was then employed in an affinity chromatography approach that resulted in the identification of the protein 14-3-3zeta as having selective affinity for the chromatography matrix that was derivatized with a salicylanilide that inhibited IL-12p40 production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131768
Salicylanilide, 98%
C13H11NO2