EMAIL THIS PAGE TO A FRIEND
Stem Cells and Development
Histone H3 modifications associated with differentiation and long-term culture of mesenchymal adipose stem cells.
PMID 18771397
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-942
Anti-acetyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit