EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
Improved analysis of bile acids in tissues and intestinal contents of rats using LC/ESI-MS.
PMID 18772484
Abstract
To evaluate bile acid (BA) metabolism in detail, we established a method for analyzing BA composition in various tissues and intestinal contents using ultra performance liquid chromatography/electrospray ionization mass spectrometry (UPLC/ESI-MS). Twenty-two individual BAs were determined simultaneously from extracts. We applied this method to define the differences in BA metabolism between two rat strains, WKAH and DA. The amount of total bile acids (TBAs) in the liver was significantly higher in WKAH than in DA rats. In contrast, TBA concentration in jejunal content, cecal content, colorectal content, and feces was higher in DA rats than in WKAH rats. Nearly all BAs in the liver were in the taurine- or glycine-conjugated form in DA rats, and the proportion of conjugated liver BAs was up to 75% in WKAH rats. Similar trends were observed for the conjugation rates in bile. The most abundant secondary BA in cecal content, colorectal content, and feces was hyodeoxycholic acid in WKAH rats and omega-muricholic acid in DA rats. Analyzing detailed BA profiles, including conjugation status, in a single run is possible using UPLC/ESI-MS. This method will be useful for investigating the roles of BA metabolism under physiological and pathological conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3878
Hyodeoxycholic acid, ≥98%
C24H40O4