EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effect of biocompost-amendment on degradation of triazoles fungicides in soil.
PMID 18773131
Abstract
Soil amendments play an important role in management of pesticide residues. Present study reports the effect of biocompost-amendment on degradation of penconazole and propiconazole (triazole fungicides) in a sandy loam soil under flooded and nonflooded (60% water holding capacity) conditions. Penconazole degraded at faster rate than propiconazole. Both the fungicides were found to be more persistent in flooded soil than nonflooded soil, but application of biocompost at 2.5% and 5.0% levels enhanced their degradation under both moisture regimes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36189
Penconazol, PESTANAL®, analytical standard
C13H15Cl2N3