EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series (2004)
Development of a novel selective detection method for 5-formyl-2'-deoxyuridine by fluorescent derivatization.
PMID 18776362
Abstract
5-Formyl-2'-deoxyuridine (fdUrd) is derived from the oxidation of thymidine and is known to induce mutation (A:T to G:C) in DNA. We planned to develop a novel selective detection method for fdUrd in oligodeoxynucleotide (ODN) based on specific fluorescent derivatization of fdUrd using 2-aminothiophenol derivatives (ATs) as fluorogenic reagents. To achieve this goal, we first synthesized 5-(benzothiazol-2-yl)-2'-deoxyuridine derivatives (btdUrds) via the reaction between fdUrd and 5-substituted ATs. We report herein the synthesis and UV absorbance and fluorescence properties of btdUrds in different pH conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274240
2-Aminothiophenol, 99%
C6H7NS
123137
2-Aminothiophenol, technical grade, 90%
C6H7NS