EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in chemistry : MRC
Covalent bonding of azoles to quaternary protoberberine alkaloids.
PMID 18781707
Abstract
Adducts of the quaternary protoberberine alkaloids (QPA) berberine, palmatine, and coptisine were prepared with nucleophiles derived from pyrrole, pyrazole, imidazole, and 1,2,4-triazole. The products, 8-substituted 7,8-dihydroprotoberberines, were identified by mass spectrometry and 1D and 2D NMR spectroscopy, including (1)H--(15)N shift correlations at natural abundance. In addition, two adducts of QPA with chloroform and methanethiolate were characterized by using NMR data. Single-crystal X-ray structures of 8-pyrrolyl-7,8-dihydroberberine, 8-pyrazolyl-7,8-dihydroberberine, and 8-imidazolyl-7,8-dihydroberberine are also presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
361615
Palmatine chloride hydrate, 97%
C21H22ClNO4 · xH2O