EMAIL THIS PAGE TO A FRIEND
Protein and peptide letters
Structural and spectroscopic elucidation of tetrapetide glycyl-(L)-prolyl-glycyl-glycine and its hydrogensquarate.
PMID 18782067
Abstract
The IR-spectroscopic and structural elucidation of tetrapeptide glycyl-(L)-prolyl-glycyl-glycine and its hydrogensquarate was performed by employing linear-polarized IR-spectroscopy of oriented colloid suspensions in nematic host as well as mass spectrometry. Quantum-chemical ab initio calculations were carried out in order to evaluate both the electronic structure and optical properties of the compound studied.