EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Quantitative evaluation of PEPT1 contribution to oral absorption of cephalexin in rats.
PMID 18784906
Abstract
PEPT1 mediates the intestinal absorption of many drugs, but its contribution to oral absorption of drugs is still controversial. The objective of this study is to quantitatively evaluate the contribution of PEPT1 to oral absorption of cephalexin, a typical substrate for PEPT1, in rats. The absorbability of cephalexin via PEPT1 or passive diffusion was assessed in five intestinal segments by utilizing glycyl-proline as a competitive inhibitor by in-situ closed loop method. Absorption kinetics of cephalexin after oral administration was predicted by GI-Transit-Absorption model. Absorbability of cephalexin was segment-dependent, and concentration-dependent in all the segments except for the lower ileum. Intrinsic absorption rate constant via PEPT1 ranged from 0.64 to 4.07 h(-1). The absorption rate constants via passive diffusion ranged from 0.78 to 1.24 h(-1). Plasma concentration-time profile of cephalexin was successfully predicted and the substantial contribution of PEPT1 to the oral absorption was calculated to be from 46% to 60% of total absorption. Simulation study indicated that 83% bioavailability would be expected for cephalexin even though PEPT1 does not function. PEPT1 substantially contributes to oral absorption of cephalexin, around a half of total absorption. However, the function of PEPT1 can be compensated by passive diffusion for cephalexin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3002
Gly-Pro
C7H12N2O3