EMAIL THIS PAGE TO A FRIEND
Journal of applied toxicology : JAT
Use of proliferation tests to evaluate the effects of complexing agents on beryllium toxicity.
PMID 18785683
Abstract
Occupational exposure to beryllium may cause chronic beryllium disease (CBD), a granulomatous interstitial pneumonitis caused by a cell-mediated immune response with delayed hypersensitivity initiated by an electrostatic interaction with the MHC class II human leukocyte antigen (HLA). Increased research efforts focus on the development of a CBD treatment by chelation therapy. This work presents an in vitro evaluation of the beneficial effects of beryllium chelation with different organic substrates. We have used a standard beryllium lymphocyte proliferation test (BeLPT) adapted for mouse splenocytes. Three complexing agents, 4,5-dihydroxy-1,3-benzenedisulfonic acid (tiron), nitrilotripropionic acid (NTP) and nitrilotriacetic acid (NTA), were tested using different protocols of the splenocyte proliferation test (SPT). We studied their corrective effect (beryllium pre-exposed splenocytes), their protective effect (ligand pre-exposed splenocytes) and their combined effects at fixed Be:L ratio of 1:2, at fixed Be concentration and at fixed L concentration. We also studied the effect of tiron in preventing splenocyte sensitization to beryllium. All three complexing agents showed a corrective effect and proved efficient in the combined effects, except NTA in the fixed Be:L ratio. Only NTP and tiron showed a significant protection at lower beryllium concentrations, while NTA was not significant. Splenocytes pre-exposed to chelated beryllium did not show sensitization while splenocytes pre-exposed to beryllium were sensitized. We observed a strong correlation between the efficiency of the complexing agent and its affinity towards beryllium. Both tiron and NTP showed a similar affinity towards the beryllium ion that is 10(7) higher than that of NTA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14270
Beryllium sulfate tetrahydrate, purum p.a., ≥99.0% (T)
BeO4S · 4H2O